missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 5/CD49e Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
214.00 € - 488.00 €
Spezifikation
| Antigen | Integrin alpha 5/CD49e |
|---|---|
| Klon | 238307 |
| Konzentration | LYOPH |
| Verdünnung | Flow Cytometry 0.25 ug/10^6 cells, Adhesion Activation, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Anwendungen | Flow Cytometry, Adhesion Activation, Immunocytochemistry, CyTOF |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18736078
![]() |
R&D Systems
MAB1864-SP |
25 μg |
214.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18122891
![]() |
R&D Systems
MAB1864 |
100 μg |
488.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Integrin alpha 5/CD49e Monoclonal specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for Flow Cytometry, Adhesion Activation, Immunocytochemistry, CyTOF-ready.Spezifikation
| Integrin alpha 5/CD49e | |
| LYOPH | |
| Flow Cytometry, Adhesion Activation, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08648 | |
| 3678 | |
| Mouse myeloma cell line NS0-derived recombinant human Integrin alpha 5/CD49e Phe42-Tyr995 Accession # P08648 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Integrin alpha 5/CD49e. |
| 238307 | |
| Flow Cytometry 0.25 ug/10^6 cells, Adhesion Activation, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5, very late activation protein 5, alpha subunit, VLA-5, VLA-5 alpha | |
| ITGA5 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title