Learn More
Abnova™ Human JRK Full-length ORF (AAH43351.1, 1 a.a. - 568 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008629-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is the human homolog of the mouse jerky gene. The encoded protein has similarity to several nuclear regulatory proteins, including centromere protein B, suggesting that it might function as a DNA-binding protein. Insertional inactivation of this gene in transgenic mice resulted in epileptic seizures. Childhood Absence Epilepsy (CAE) has been mapped to the same chromosomal location (8q24.3) as this gene, making this gene a strong candidate for CAE. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MASKPAAGKSRGEKRKRVVLTLKEKIDICTRLEKGESRKALMQEYNVGMSTLYDIRAHKAQLLRFFASSDSNKALEQRRTLHTPKLEHLDRVLYEWFLGKRSEGVPVSGPMLIEKAKDFYEQMQLTEPCVFSGGWLWRFKARHGIKKLDASSEKQSADHQAAEQFCAFFRSLAAEHGLSAEQVYNADETGLFWRCLPNPTPEGGAVPGPKQGKDRLTVLMCANATGSHRLKPLAIGKCSGPRAFKGIQHLPVAYKAQGNAWVDKEIFSDWFHHIFVPSVREHFRTIGLPEDSKAVLLLDSSRAHPQEAELVSSNVFTIFLPASVASLVQPMEQGIRRDFMRNFINPPVPLQGPHARYNMNDAIFSVACAWNAVPSHVFRRAWRKLWPSVAFAEGSSSEEELEAECFPVKPHNKSFAHILELVKEGSSCPGQLRQRQAASWGVAGREAEGGRPPAATSPAEVVWSSEKTPKADQDGRGDPGEGEEVAWEQAAVAFDAVLRFAERQPCFSAQEVGQLRALRAVFRSQQQVRRRRGALGAVVKVEALQEGPGGCGATAQSPLPCSSTAGDNSpezifikation
AAH43351.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686C24207/FLJ45729/JH8 | |
JRK | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8629 | |
JRK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASKPAAGKSRGEKRKRVVLTLKEKIDICTRLEKGESRKALMQEYNVGMSTLYDIRAHKAQLLRFFASSDSNKALEQRRTLHTPKLEHLDRVLYEWFLGKRSEGVPVSGPMLIEKAKDFYEQMQLTEPCVFSGGWLWRFKARHGIKKLDASSEKQSADHQAAEQFCAFFRSLAAEHGLSAEQVYNADETGLFWRCLPNPTPEGGAVPGPKQGKDRLTVLMCANATGSHRLKPLAIGKCSGPRAFKGIQHLPVAYKAQGNAWVDKEIFSDWFHHIFVPSVREHFRTIGLPEDSKAVLLLDSSRAHPQEAELVSSNVFTIFLPASVASLVQPMEQGIRRDFMRNFINPPVPLQGPHARYNMNDAIFSVACAWNAVPSHVFRRAWRKLWPSVAFAEGSSSEEELEAECFPVKPHNKSFAHILELVKEGSSCPGQLRQRQAASWGVAGREAEGGRPPAATSPAEVVWSSEKTPKADQDGRGDPGEGEEVAWEQAAVAFDAVLRFAERQPCFSAQEVGQLRALRAVFRSQQQVRRRRGALGAVVKVEALQEGPGGCGATAQSPLPCSSTAGDN | |
RUO | |
JRK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |