Learn More
Abnova™ Human KRTAP1-5 Full-length ORF (NP_114163.1, 1 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00083895-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the high sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21. [provided by RefSeq]
Sequence: MTCCQTSFCGYPSFSISGTCGSSCCQPSCCETSCCQPRSCQTSFCGFPSFSTSGTCSSSCCQPSCCETSCCQPSCCETSCCQPSCCQISSCGTGCGIGGGISYGQEGSSGAVSTRIRWCRPDSRVEGTYLPPCCVVSCTPPSCCQLHHAQASCCRPSYCGQSCCRPVCCCEPTCSpezifikation
NP_114163.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KAP1.5/KRTAP1.5/MGC126604 | |
KRTAP1-5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
83895 | |
KRTAP1-5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTCCQTSFCGYPSFSISGTCGSSCCQPSCCETSCCQPRSCQTSFCGFPSFSTSGTCSSSCCQPSCCETSCCQPSCCETSCCQPSCCQISSCGTGCGIGGGISYGQEGSSGAVSTRIRWCRPDSRVEGTYLPPCCVVSCTPPSCCQLHHAQASCCRPSYCGQSCCRPVCCCEPTC | |
RUO | |
KRTAP1-5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |