Learn More
Abnova™ Human LAT2 Full-length ORF (NP_054865.2, 1 a.a. - 243 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007462-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is one of the contiguous genes at 7q11.23 commonly deleted in Williams syndrome, a multisystem developmental disorder. This gene consists of at least 14 exons, and its alternative splicing generates 3 transcript variants, all encoding the same protein. [provided by RefSeq]
Sequence: MSSGTELLWPGAALLVLLGVAASLCVRCSRPGAKRSEKIYQQRSLREDQQSFTGSRTYSLVGQAWPGPLADMAPTRKDKLLQFYPSLEDPASSRYQNFSKGSRHGSEEAYIDPIAMEYYNWGRFSKPPEDDDANSYENVLICKQKTTETGAQQEGIGGLCRGDLSLSLALKTGPTSGLCPSASPEEDEESEDYQNSASIHQWRESRKVMGQLQREASPGPVGSPDEEDGEPDYVNGEVAATEASpezifikation
NP_054865.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSPC046/LAB/NTAL/WBSCR15/WBSCR5/WSCR5 | |
LAT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7462 | |
LAT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSGTELLWPGAALLVLLGVAASLCVRCSRPGAKRSEKIYQQRSLREDQQSFTGSRTYSLVGQAWPGPLADMAPTRKDKLLQFYPSLEDPASSRYQNFSKGSRHGSEEAYIDPIAMEYYNWGRFSKPPEDDDANSYENVLICKQKTTETGAQQEGIGGLCRGDLSLSLALKTGPTSGLCPSASPEEDEESEDYQNSASIHQWRESRKVMGQLQREASPGPVGSPDEEDGEPDYVNGEVAATEA | |
RUO | |
LAT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |