Learn More
STEMCELL Technologies Human Latent TGF-beta 1 ELISA Kit
Artikelnummer. 17118031
Encompass_Preferred
Click to view available options
Menge:
2 Platten
Artikelnummer. 17118031
Marke: STEMCELL Technologies 02018
om dit product te kopen Registreer vandaag om een webaccount aan te maken
For detection and measurement of human latent transforming growth factor-beta 1
  • The Human Latent Transforming Growth Factor-beta 1 (TGF-β1) Kit is designed for the quantitative detection and measurement of latent human TGF-β1 in biological fluids such as plasma and cell culture supernatants
  • TGF-β1 stimulates cells of mesenchymal origin, but inhibits cells of epithelial or neuroectodermal origin
  • TGF-β1 is secreted in a latent, biologically inactive form and associates with Latency Associated Protein (LAP) to form a latent complex
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
  • The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Spezifikation
Beschreibung Dendritic Cells; Macrophages; Platelets; T Cells; Tumor Cells
Menge 2 Platten
Berichtigung von Produktinhalten
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.