Learn More
ACROBiosystems Human LILRB2 / CD85d / ILT4 Protein, Fc tag
Marke:  ACROBiosystems CDD-H5259-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15896377
  • 1881.77 € / 1 Milligramm
Verfügbar ab: 05-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Leukocyte immunoglobulin-like receptor subfamily B member 2 (LILRB2) is also known as CD85 antigen-like family member D (CD85d), Immunoglobulin-like transcript 4 (ILT-4), Monocyte / macrophage immunoglobulin-like receptor 10 (MIR-10), which is a member of the the subfamily B class of LIR receptors. LILRB2 is receptor for class I MHC antigens. LILRB2 recognizes a broad spectrum of HLA-A, HLA-B, HLA-C and HLA-G alleles. LILRB2 competes with CD8A for binding to class I MHC antigens. LILRB2 / CD85d inhibits FCGR1A-mediated phosphorylation of cellular proteins and mobilization of intracellular calcium ions.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 74.4 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH36827
74.4 kDa
1 mg
Gln 22 - Val 461
LILRB2,ILT4,LIR2,MIR10,CD85d
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Human
Powder
>90% as determined by SDS-PAGE
Human LILRB2, Fc Tag (CDD-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 461 (Accession # AAH36827).
-20°C
LILRB2
Fc Tag
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins