Learn More
Abnova™ Human MUC7 Partial ORF (NP_689504, 36 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004589-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a small salivary mucin, which is thought to play a role in facilitating the clearance of bacteria in the oral cavity and to aid in mastication, speech, and swallowing. The central domain of this glycoprotein contains tandem repeats, each composed of 23 amino acids. The most common allele contains 6 repeats, and some alleles may be associated with susceptibility to asthma. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene
Sequence: HQSPKSHFELPHYPGLLAHQKPFIRKSYKCLHKRCRPKLPPSPNNPPKFPNPHQPPKHPDKNSSVVNPTLVATTQIPSVTFPSASTKITTLPNVTFLPQNSpecifica
NP_689504 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HQSPKSHFELPHYPGLLAHQKPFIRKSYKCLHKRCRPKLPPSPNNPPKFPNPHQPPKHPDKNSSVVNPTLVATTQIPSVTFPSASTKITTLPNVTFLPQN | |
RUO | |
MUC7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4589 | |
MUC7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686J03256/FLJ27047/MG2/MGC34772 | |
MUC7 | |
Recombinant | |
wheat germ expression system |