Learn More
ACROBiosystems Human Nectin-2 / CD112 Protein, His Tag
Marke:  ACROBiosystems PV2-H52E2-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15887567
  • 2258.12 € / 1 Milligramm
Verfügbar ab: 04-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Poliovirus receptor-related 2 (PVRL2) is also known as nectin-2 and CD112 (formerly herpesvirus entry mediator B, HVEB), which is a human plasma membrane glycoprotein. PVRL2 is one of the plasma membrane components of adherens junctions. PVRL2 also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in PVRL2 gene have been associated with differences in the severity of multiple sclerosis. Also, PVRL2 binds with low affinity to TIGIT. Furthermore, PVRL2 acts as a receptor for herpes simplex virus 1 (HHV-1) mutant Rid1, herpes simplex virus 1 (HHV-2) and pseudorabies virus (PRV).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.4 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_002847.1
37.4 kDa
1 mg
Gln 32 - Leu 360
CD112,Nectin-2,PVRL2
Unconjugated
PBS, pH7.4
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Human Nectin-2, His Tag (PV2-H52E2) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).
-20°C
Nectin-2
His Tag
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins