Learn More
ACROBiosystems Human Neprilysin / MME / CD10 Protein, Tag Free
483.87 € - 2951.55 €
Spezifikation
Zugriffsnummer | NP_000893.2 |
---|---|
Molekulargewicht | 80.0 kDa |
Quelle | Human Neprilysin, Tag Free (MME-H521b) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession # NP_000893.2). |
Immunogen | Tyr 52 - Trp 750 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15807317
|
ACROBiosystems
MME-H521B-1MG |
1 mg |
2951.55 €
1 Milligramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15817317
|
ACROBiosystems
MME-H521B-50UG |
50 μg |
483.87 €
50 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Cluster of differentiation 10 (CD10) is also known as membrane metallo-endopeptidase, neutral endopeptidase (NEP), Neprilysin, and common acute lymphoblastic leukemia antigen (CALLA), is a 90-110-kDa type II transmembrane glycoprotein normally expressed by a variety of tissues, including epithelial cells of the prostate, kidney, intestine, endometrium, adrenal glands, and lung. This zinc-dependent metalloprotease enzyme cleaves peptide bonds on the amino side of hydrophobic residues and inactivates a variety of physiologically active secreted peptides. CD20 is thought to be the rate-limiting degrading enzyme of amyloid β peptide (Aβ) whose abnormal misfolding and aggregation in neural tissue has been implicated in the development of Alzheimer's disease (AD). CD10 is also identified as the common acute lymphoblastic leukemia antigen (CALLA) present on leukemic cells of pre-B phenotype, and thus serves as the most important biomarker in the diagnosis of human acute lymphocytic leukemia (ALL).
This protein carries no "e;tag"e;. The protein has a calculated MW of 80.0 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000893.2 | |
Human Neprilysin, Tag Free (MME-H521b) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession # NP_000893.2). | |
-20°C | |
Neprilysin | |
Human | |
20 mM Tris, 100 mM NaCl, pH8.0 | |
HEK293 | |
Enzymes |
80.0 kDa | |
Tyr 52 - Trp 750 | |
MME,CALLA,CD10,DKFZp686O16152,MGC126681,MGC126707,NEP,SFE,Neprilysin | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |