Learn More
ACROBiosystems Human Neuregulin-4 / NRG4 Protein, His Tag
Marke: ACROBiosystems NR4-H52H3-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Neuregulin 4 also known as NRG4 is a member of the neuregulin protein family which in humans is encoded by the NRG4 gene. Loss of expression of NRG4 is frequently seen in advanced bladder cancer while increased NRG4 expression correlates to better survival. The neuregulins, including NRG4, activate type-1 growth factor receptors (EGFR) to initiating cell-to-cell signaling through tyrosine phosphorylation. Furthermore, NRG4 is a low affinity ligand for the ERBB4 tyrosine kinase receptor. Concomitantly recruits ERBB1 and ERBB2 coreceptors, resulting in ligand-stimulated tyrosine phosphorylation and activation of the ERBB receptors. NRG4 does not bind to the ERBB1, ERBB2 and ERBB3 receptors.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 8.6 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
8.6 kDa | |
Human Neuregulin-4, His Tag (NR4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Pro 2 - Phe 62 (Accession # Q8WWG1). | |
-20°C | |
Neuregulin-4 | |
Human | |
50 mM Tris, 100 mM NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
1 mg | |
Pro 2 - Phe 62 | |
Neuregulin-4,NRG4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |