missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp30/NCR3 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
214.00 € - 497.00 €
Spezifikation
| Antigen | NKp30/NCR3 |
|---|---|
| Klon | 210845 |
| Verdünnung | Flow Cytometry 10 uL/10^6 cells |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18733069
![]() |
R&D Systems
FAB1849P-025 |
25 Tests |
214.00 €
25 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18728576
![]() |
R&D Systems
FAB1849P-100 |
100 Tests |
497.00 €
100 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Flow Cytometry.Spezifikation
| NKp30/NCR3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 259197 | |
| Mouse T cell hybridoma transfected with human NKp30/NCR3 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 210845 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS | |
| NCR3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human NKp30/NCR3 on cell transfectants and peripheral blood NK cells. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto