Learn More
Abnova™ Human NKX3-1 Partial ORF (NP_006158, 100 a.a. - 209 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004824-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.[supplied by OMIM]
Sequence: GSYLLDSENTSGALPRLPQTPKQPQKRSRAAFSHTQVIELERKFSHQKYLSAPERAHLAKNLKLTETQVKIWFQNRRYKTKRKQLSSELGDLEKHSSLPALKEEAFSRASSpezifikation
NP_006158 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GSYLLDSENTSGALPRLPQTPKQPQKRSRAAFSHTQVIELERKFSHQKYLSAPERAHLAKNLKLTETQVKIWFQNRRYKTKRKQLSSELGDLEKHSSLPALKEEAFSRAS | |
RUO | |
NKX3-1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4824 | |
NKX3-1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAPX2/NKX3/NKX3.1/NKX3A | |
NKX3-1 | |
Recombinant | |
wheat germ expression system |