Learn More
Abnova™ Human NPTX1 Partial ORF (NP_002513.2, 141 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004884-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
NPTX1 is a member of the neuronal pentraxin gene family. Neuronal pentraxin 1 is similar to the rat NP1 gene which encodes a binding protein for the snake venom toxin taipoxin. Human NPTX1 mRNA is exclusively localized to the nervous system. [provided by RefSeq]
Sequence: KTRLENLEQYSRLNSSSQTNSLKDLLQSKIDELERQVLSRVNTLEEGKGGPRNDTEERVKIETALTSLHQRISELEKGQKDNRPGDKFQLTFPLRTNYMYSpezifikation
NP_002513.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KTRLENLEQYSRLNSSSQTNSLKDLLQSKIDELERQVLSRVNTLEEGKGGPRNDTEERVKIETALTSLHQRISELEKGQKDNRPGDKFQLTFPLRTNYMY | |
RUO | |
NPTX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4884 | |
NPTX1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686J2446/MGC105123/NP1 | |
NPTX1 | |
Recombinant | |
wheat germ expression system |