Learn More
Abnova™ Human OFD1 Full-length ORF (AAH99658.1, 1 a.a. - 550 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008481-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is located on the X chromosome and encodes a centrosomal protein. A knockout mouse model has been used to study the effect of mutations in this gene. The mouse gene is also located on the X chromosome, however, unlike the human gene it is not subject to X inactivation. Mutations in this gene are associated with oral-facial-digital syndrome type I and Simpson-Golabi-Behmel syndrome type 2. Many pseudogenes have been identified; a single pseudogene is found on chromosome 5 while as many as fifteen have been found on the Y chromosome. Alternatively spliced transcripts have been described for this gene but the biological validity of these transcripts has not been determined. [provided by RefSeq]
Sequence: MLNEKVKEMSDYSLLKEEKLELLAQNKLLKQQLEESRNENLRLLNRLAQPAPELAVFQKELRKAEKAIVVEHEEFESCRQALHKQLQDEIEHSAQLKAQILGYKASVKSLTTQVADLKLQLKQTQTALENEVYCNPKQSVIDRSVNGLINGNVVPCNGEISGDFLNNPFKQENVLARMVASRITNYPTAWVEGSSPDSDLEFVANTKARVKELQQEAERLEKAFRSYHRRVIKNSAKSPLAAKSPPSLHLLEAFKNITSSSPERHIFGEDRVVSEQPQVGTLEERNDVVEALTGSAASRLRGGTSSRRLSSTPLPKAKRSLESEMYLEGLGRSHIASPSPCPDRMPLPSPTESRHSLSIPPVSSPPEQKVGLYRRQTELQDKSEFSDVDKLAFKDNEEFESSFECVDQKQIEEQKEEEKIREQQVKERRQREERRQSNLQEVLERERRELEKLYQERKMIEESLKIKIKKELEMENELEMSNQEIKDKSAHSENPLEKYMKIIQQEQDQESADKSSKKMVQEGSLVDTLQSSDKVESLTGFSHEELDDSWSpezifikation
AAH99658.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
71-7A/CXorf5/MGC117039/MGC117040/SGBS2 | |
OFD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8481 | |
OFD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLNEKVKEMSDYSLLKEEKLELLAQNKLLKQQLEESRNENLRLLNRLAQPAPELAVFQKELRKAEKAIVVEHEEFESCRQALHKQLQDEIEHSAQLKAQILGYKASVKSLTTQVADLKLQLKQTQTALENEVYCNPKQSVIDRSVNGLINGNVVPCNGEISGDFLNNPFKQENVLARMVASRITNYPTAWVEGSSPDSDLEFVANTKARVKELQQEAERLEKAFRSYHRRVIKNSAKSPLAAKSPPSLHLLEAFKNITSSSPERHIFGEDRVVSEQPQVGTLEERNDVVEALTGSAASRLRGGTSSRRLSSTPLPKAKRSLESEMYLEGLGRSHIASPSPCPDRMPLPSPTESRHSLSIPPVSSPPEQKVGLYRRQTELQDKSEFSDVDKLAFKDNEEFESSFECVDQKQIEEQKEEEKIREQQVKERRQREERRQSNLQEVLERERRELEKLYQERKMIEESLKIKIKKELEMENELEMSNQEIKDKSAHSENPLEKYMKIIQQEQDQESADKSSKKMVQEGSLVDTLQSSDKVESLTGFSHEELDDSW | |
RUO | |
OFD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |