missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Oncostatin M/OSM Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Marke: R&D Systems MAB295-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Spezifikation
| Oncostatin M/OSM | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC20461, oncostatin M, oncostatin-M | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 5008 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 17001 | |
| Western Blot 1 ug/mL, Neutralization 0.5-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P13725 | |
| OSM | |
| E. coli-derived recombinant human Oncostatin M/OSM Ala26-Arg234 Accession # P13725 | |
| 100 μg | |
| Primary | |
| Detects human Oncostatin M in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) IL-6, rhIL-11, rhCNTF, or rhLIF. | |
| Human | |
| Ascites |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur