Learn More
Human Phospho-MLKL (T357) Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB9187-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MLKL Monoclonal specifically detects MLKL in Human samples. It is validated for Immunohistochemistry.
Spezifikation
| MLKL | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
| Phosphopeptide containing the human MLKL T357 site Accession # Q8NB16 | |
| 100 μg | |
| Primary | |
| Detects human MLKL when phosphorylated at T357. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 954702 | |
| Immunohistochemistry 1-25 ug/mL | |
| Q8NB16 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 197259 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |