missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human PLGF / PGF Protein, His Tag
350.80 € - 2153.18 €
Spezifikation
Zugriffsnummer | NP_002623.2 |
---|---|
Molekulargewicht | 18.2 kDa |
Quelle | Human PLGF, His Tag (PGF-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession # NP_002623.2). |
Immunogen | Leu 19 - Arg 170 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15847537
|
ACROBiosystems
PGF-H5229-500UG |
500 μg |
2153.18 €
500 Mikrogramm |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15837537
|
ACROBiosystems
PGF-H5229-20UG |
20 μg |
350.80 €
20 Mikrogramm |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Placental growth factor (PGF) is also known as vascular endothelial growth factor-related protein, PLGF and PlGF2, is a member of the VEGF (vascular endothelial growth factor) sub-family - a key molecule in angiogenesis and vasculogenesis, in particular during embryogenesis. The main source of PGF during pregnancy is the placental trophoblast. PGF is also expressed in many other tissues, including the villous trophoblast. PGF is actived in angiogenesis and endothelial cell growth, stimulating their proliferation and migration. PlGF2 binds NRP1/neuropilin-1 and NRP2/neuropilin-2 in a heparin-dependent manner. Also promotes cell tumor growth.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.2 kDa. The protein migrates as 28-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002623.2 | |
Human PLGF, His Tag (PGF-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession # NP_002623.2). | |
-20°C | |
PLGF | |
Human | |
100 mM Acetic Acid, pH3.0 | |
HEK293 | |
Bio-Markers and CD Antigens |
18.2 kDa | |
Leu 19 - Arg 170 | |
PGF,PLGF,PlGF,PGFL,SHGC-10760 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |