missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro-Cathepsin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 537.00 €
Spezifikation
| Antigen | Cathepsin B |
|---|---|
| Klon | 155714 |
| Verdünnung | Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Anwendungen | Immunoprecipitation, ELISA |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18799398
![]() |
R&D Systems
MAB2176-SP |
25 μg |
209.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18190911
![]() |
R&D Systems
MAB2176 |
500 μg |
537.00 €
500 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Cathepsin B Monoclonal specifically detects Cathepsin B in Human samples. It is validated for Immunoprecipitation, ELISA Capture (Matched Antibody Pair), ELISA Capture (Matched Antibody Pair).Spezifikation
| Cathepsin B | |
| Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, CTSB, cysteine protease, EC 3.4.22, EC 3.4.22.1 | |
| CTSB | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 155714 | |
| Immunoprecipitation, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P07858 | |
| 1508 | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin B Arg18-Ile339 Accession # P07858 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Cathepsin B. Does not cross-react with recombinant human Cathepsin A, C, D, E, L, S, V, X/Z/P, SA, or recombinant mouse Cathepsin B. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts