Learn More
ACROBiosystems Human Prolactin / PRL Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marke: ACROBiosystems PRN-H5257-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Prolactin (gene name PRL) is a secreted neuroendocrine pituitary hormone that acts primarily on the mammary gland to promote lactation, but has pleiotropic effects in both males and females. Prolactin is synthesized as a prohormone. Following cleavage of the signal peptide, the length of the mature hormone is between 194 and 199 amino acids, depending on species. Hormone structure is stabilized by three intramolecular disulfide bonds. Excessive secretion of prolactin - hyperprolactinemia - is a relative common disorder in humans. This condition has numerous causes, including prolactin-secreting tumors and therapy with certain drugs. The prolactin receptor (gene name PRLR) is a transmembrane type I glycoprotein that belongs to the cytokine hematopoietic receptor family. Expression of the prolactin receptor is widespread. Each prolactin molecule is thought to bind two receptor molecules
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 49.8 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000939.1 | |
100 μg | |
Leu 29 - Cys 227 | |
PRL,Prolactin | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
49.8 kDa | |
Human Prolactin, Mouse IgG2a Fc Tag, low endotoxin (PRN-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Accession # Q5THQ0-1). | |
-20°C | |
Prolactin | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Cytokines and Growth Factors |