missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Properdin Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | NP_001138724.1 |
---|---|
Molekulargewicht | 50.7 kDa |
Quelle | Human Properdin, His Tag (PRN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession # P27918-1). |
Immunogen | Asp 28 - Leu 469 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16497969
|
ACROBiosystems
PRN-H52H3-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16407979
|
ACROBiosystems
PRN-H52H3-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Properdin is also known as Complement factor P, CFP, PFC, a positive regulator of the alternate pathway of complement. It binds to and stabilizes the C3- and C5-convertase enzyme complexes. Properdin is produced by many different leukocyte subsets and circulates as cyclic oligomers of monomeric subunits. Properdin on platelet/granulocyte aggregates(PGA) formation are tightly regulated by Factor H, properdin enhances PGA formation via increased production of C5a, and that inhibition of properdin function has therapeutic potential to limit thromboinflammation in diseases characterized by increased PGA formation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.7 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001138724.1 | |
Human Properdin, His Tag (PRN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession # P27918-1). | |
-20°C | |
Properdin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
50.7 kDa | |
Asp 28 - Leu 469 | |
Properdin,Complement factor P,CFP,PFC | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |