missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PSG6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB85983-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PSG6 Monoclonal specifically detects PSG6 in Human samples. It is validated for ELISA.
Spezifikation
| PSG6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CGM3, pregnancy specific beta-1-glycoprotein 6, Pregnancy-specific beta-1-glycoprotein 10, Pregnancy-specific beta-1-glycoprotein 12, pregnancy-specific beta-1-glycoprotein 6, Pregnancy-specific glycoprotein 10, Pregnancy-specific glycoprotein 12, Pregnancy-specific glycoprotein 6, PS-beta-G-10, PS-beta-G-12, PS-beta-G-6, PSBG-10, PSBG-12, PSBG-6, PSG10, PSG12, PSG-6, PSGGB | |
| Human embryonic kidney cell line HEK-293-derived transfected with human PSG6 Met1-Gly424 Accession # Q00889 | |
| 25 μg | |
| Primary | |
| Detects human PSG6 in direct ELISAs. | |
| Human | |
| Purified |
| ELISA | |
| 935947 | |
| ELISA | |
| Q00889 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5675 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
RUO
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur