missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PSG6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
196.00 € - 407.00 €
Spezifikation
| Antigen | PSG6 |
|---|---|
| Klon | 935947 |
| Verdünnung | ELISA |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18758918
![]() |
R&D Systems
MAB85983-SP |
25 μg |
196.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18744518
![]() |
R&D Systems
MAB85983-100 |
100 μg |
407.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Description
PSG6 Monoclonal specifically detects PSG6 in Human samples. It is validated for ELISA.Specifications
| PSG6 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CGM3, pregnancy specific beta-1-glycoprotein 6, Pregnancy-specific beta-1-glycoprotein 10, Pregnancy-specific beta-1-glycoprotein 12, pregnancy-specific beta-1-glycoprotein 6, Pregnancy-specific glycoprotein 10, Pregnancy-specific glycoprotein 12, Pregnancy-specific glycoprotein 6, PS-beta-G-10, PS-beta-G-12, PS-beta-G-6, PSBG-10, PSBG-12, PSBG-6, PSG10, PSG12, PSG-6, PSGGB | |
| Human embryonic kidney cell line HEK-293-derived transfected with human PSG6 Met1-Gly424 Accession # Q00889 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PSG6 in direct ELISAs. |
| 935947 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q00889 | |
| 5675 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title