Learn More
Abnova™ Human PVRL3 Partial ORF (NP_056295, 59 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00025945-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Nectins (e.g., PVRL1; MIM 600644) are immunoglobulin-like adhesion molecules that interact with afadin (AF6; MIM 159559). Afadin is an actin filament-binding protein that connects nectins to the actin cytoskeleton. The nectin-afadin system organizes adherens junctions cooperatively with the cadherin (see MIM 192090)-catenin (see MIM 116805) system in epithelial cells.[supplied by OMIM]
Sequence: PIIVEPHVTAVWGKNVSLKCLIEVNETITQISWEKIHGKSSQTVAVHHPQYGFSVQGEYQGRVLFKNYSLNDATITLHNIGFSDSGKYICKAVTFPLGNAQSSTTVTVSpezifikation
NP_056295 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PIIVEPHVTAVWGKNVSLKCLIEVNETITQISWEKIHGKSSQTVAVHHPQYGFSVQGEYQGRVLFKNYSLNDATITLHNIGFSDSGKYICKAVTFPLGNAQSSTTVTV | |
RUO | |
PVRL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25945 | |
PVRL3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD113/CDw113/DKFZp566B0846/FLJ90624/PPR3/PRR3/PVRR3/nectin-3 | |
PVRL3 | |
Recombinant | |
wheat germ expression system |