Learn More
STEMCELL Technologies Human Recombinant BDNF, ACF
Beschreibung
- Brain-derived neurotrophic factor (BDNF), like nerve growth factor (NGF), neurotrophin-3 (NT-3), and neurotrophin-4 (NT-4), is a member of the NGF family of neurotrophins, which are required for the differentiation and survival of specific neuronal subpopulations in both the central and the peripheral nervous systems
- BDNF binds with high affinity to the TRKB kinase receptor, and activates AKT and ERK pathways
- It is expressed in hippocampus, cortex, and synapses of the basal forebrain
- BDNF acts as a survival factor for human embryonic stem cells when plated on either feeder cells or Corning™ Matrigel™
Spezifikation
Spezifikation
| Name | Abrineurin, ANON2, BULN2, Neurotrophin, MGC34632 |
| Menge | 10 μg |
| Zelltyp | Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons |
| Reinheits- oder Qualitätsgrad | ≥95% |