Learn More
STEMCELL Technologies Human Recombinant HGF
Click to view available options
Menge:
10 μg
1000 μg
50 μg
Packungsgröße:
50 Mikrogramm
Beschreibung
- Hepatocyte growth factor/scatter factor (HGF/SF) is a pleiotropic growth factor that promotes proliferation, motility, survival, and differentiation
- HGF binds and promotes the dimerization and activation of the receptor tyrosine kinase c-MET, and stimulates PI3K/AKT, FAK, JNK, and ERK1/2 signaling (Organ & Tsao)
- HGF stimulates migration of cells during embryogenesis, induces cell motility and scattering of epithelial cells, and regulates epithelial-mesenchymal transition
- It is a chemoattractant for motor neurons, and regulates the development of sensory, sympathetic, parasympathetic, and cortical neurons as well as the proliferation of oligodendrocytes and glial development
- Additionally, HGF is cytoprotective and regulates liver growth and regeneration (Nakamura et al.)
Spezifikation
Spezifikation
| Name | DFNB39, F-TCF, Fibroblast-derived tumor cytotoxic factor, Hepapoietin A, HPTA, Lung fibroblast-derived mitogen, Scatter factor, SF |
| Menge | 10 μg |
| Zelltyp | Airway Cells; Hepatic Cells; Intestinal Cells; Mammary Cells; Neurons; Pluripotent Stem Cells; Prostate Cells |
| Reinheits- oder Qualitätsgrad | ≥95% |