Learn More
STEMCELL Technologies Human Recombinant PDGF-DD
Artikelnummer. 17139891
Encompass_Preferred
Click to view available options
Menge:
10 μg
50 μg
Unit Size:
50 Mikrogramm
This item is not returnable. View return policy
Product Code. 17139891
Brand: STEMCELL Technologies 78222
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Platelet-derived growth factor DD
  • The platelet-derived growth factor (PDGF) family has five heparin-binding members that assemble into four homodimers (PDGF-AA, PDGF-BB, PDGF-CC, and PDGF-DD) and one heterodimer (PDGF-AB; Fretto et al.; Li & Eriksson)
  • PDGF signals through the receptor tyrosine kinases PDGFRα and PDGFRβ
  • It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al.) PDGF is a potent mitogen for cells of mesenchymal origin, such as fibroblasts, glial cells, and vascular smooth muscle cells
  • PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.) PDGF-DD promotes growth and survival of renal artery smooth muscle cells and lens epithelial cells, and can act as a macrophage chemoattractant (Changsirikulchai et al.; Lokker et al.; Ray et al.; Uutela et al.)
Specifications
Name IEGF, Platelet-derived growth factor D, SCDGFB
Menge 10 μg
Zelltyp Hematopoietic Stem and Progenitor Cells; Mesenchymal Cells, PSC-Derived; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Other; Pluripotent Stem Cells
Reinheits- oder Qualitätsgrad ≥95%