Learn More
STEMCELL Technologies Human Recombinant PDGF-DD
Beschreibung
- The platelet-derived growth factor (PDGF) family has five heparin-binding members that assemble into four homodimers (PDGF-AA, PDGF-BB, PDGF-CC, and PDGF-DD) and one heterodimer (PDGF-AB; Fretto et al.; Li & Eriksson)
- PDGF signals through the receptor tyrosine kinases PDGFRα and PDGFRβ
- It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al.) PDGF is a potent mitogen for cells of mesenchymal origin, such as fibroblasts, glial cells, and vascular smooth muscle cells
- PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.) PDGF-DD promotes growth and survival of renal artery smooth muscle cells and lens epithelial cells, and can act as a macrophage chemoattractant (Changsirikulchai et al.; Lokker et al.; Ray et al.; Uutela et al.)
Spezifikation
Spezifikation
| Name | IEGF, Platelet-derived growth factor D, SCDGFB |
| Menge | 10 μg |
| Zelltyp | Hematopoietic Stem and Progenitor Cells; Mesenchymal Cells, PSC-Derived; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Other; Pluripotent Stem Cells |
| Reinheits- oder Qualitätsgrad | ≥95% |