Learn More
STEMCELL Technologies Human Recombinant RANKL
Artikelnummer. 17119861
Encompass_Preferred
Click to view available options
Menge:
10 μg
100 μg
1000 μg
Packungsgröße:
10 Mikrogramm
Artikelnummer. 17119861
Marke: STEMCELL Technologies 78214
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Receptor activator of nuclear factor kappa-B ligand
  • Receptor activator of nuclear factor kappa-B ligand (RANKL) is a member of the tumor necrosis factor (TNF) superfamily (Anderson et al.) Cytokines in the TNF superfamily are involved in a variety of long-term cellular activities such as differentiation, proliferation, and cell death (MacEwan)
  • RANKL is a type II homotrimeric transmembrane protein expressed in both a membrane-bound and secreted form (Ikeda et al.) RANKL binds to the receptor activator of nuclear factor kappa-B (RANK)
  • Upon binding to its receptor, RANKL activates the AKT signaling pathway (Moon et al.) Osteoprotegerin (OPG) may also bind RANKL, and this binding competes with RANKL-RANK binding (Lacey et al.) RANKL is involved in osteoclastogenesis (Lacey et al.; Yasuda et al.) and T cell activation (Wong et al.)
Spezifikation
Name CD254, hRANKL2, ODF, OPGL, OPTB2, Osteoclast differentiation factor, soluble Receptor activator of NF-κB ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TNF superfamily member 11, TNLG6B, Tumor necrosis factor superfamily member 11, TRAN
Menge 10 μg
Zelltyp Airway Cells; Cancer Cells and Cell Lines; Dendritic Cells; Intestinal Cells; Mammary Cells; Osteoblasts; Other; Pancreatic Cells; Prostate Cells; T Cells
Reinheits- oder Qualitätsgrad ≥95%