Learn More
ACROBiosystems Human S100A10 Protein, His Tag
362.89 € - 2419.31 €
Spezifikation
Zugriffsnummer | AAH15973 |
---|---|
Molekulargewicht | 12.0 kDa |
Quelle | Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession # AAH15973). |
Immunogen | Met 1 - Lys 97 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15887627
|
ACROBiosystems
S10-H5127-100UG |
100 μg |
362.89 €
100 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15897627
|
ACROBiosystems
S10-H5127-1MG |
1 mg |
2419.31 €
1 Milligramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
S100 calcium-binding protein A10 (S100A10), a member of the S100 family of proteins containing two EF-hand calcium-binding motifs, is also known as protein S100-A10, calpactin I light chain, cellular ligand of annexin II, p10 protein and p11, which can be found as a free monomer, a homodimer, or a heterotetramer composed of a p11 dimer complex with two molecules of annexin II. S100A10 is an integral part of cellular structural scaffolding that interacts with plasma membrane proteins through its association with annexin II. S100A10 also works together with cytosolic and peripheral membrane-associated proteins such as AHNAK in the development of the intracellular membrane. Furthermore,S100A10 has been implicated in the transportation of proteins involved in mood regulation, nociception, and cell polarization. Complexed with the annexin II, S100A10 binds receptor and channel proteins and guides them to the cell surface, resulting in increased membrane localization and consequent magnified functional expression of these proteins.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.0 kDa. The protein migrates as 14 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH15973 | |
Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession # AAH15973). | |
-20°C | |
S100A10 | |
Human | |
50 mM Tris, 150 mM NaCl, pH8.0 | |
E.coli | |
Other Recombinant Proteins |
12.0 kDa | |
Met 1 - Lys 97 | |
S100A10,p11,p10,ANX2LG,CAL1L,CLP11 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |