Learn More
ACROBiosystems Human S100B Protein, His Tag
Marke: ACROBiosystems S1B-H5143-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
S-100 protein beta chain (S100B),a member of the S-100 family, is also known as S-100 protein subunit beta and S100 calcium-binding protein B, which contains two EF-hand domains. S100B can weakly bind calcium but bind zinc very tightly-distinct binding sites with different affinities exist for both ions on each monomer. Also, S100B can bind to and initiate the activation of STK38 by releasing autoinhibitory intramolecular interactions within the kinase. Interaction with AGER after myocardial infarction,S100B may play a role in myocyte apoptosis by activating ERK1/2 and p53/TP53 signaling. Fuethermore,S100B could assist ATAD3A cytoplasmic processing, preventing aggregation and favoring mitochondrial localization. S100B may mediate calcium-dependent regulation on many physiological processes by interacting with other proteins, such as TPR-containing proteins, and modulating their activity.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 11.5 kDa. The protein migrates as 11 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_006263 | |
100 μg | |
Ser 2 - Glu 92 | |
S100B,NEF,S100,S100beta | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
11.5 kDa | |
Human S100B, His Tag (S1B-H5143) is expressed from E.coli cells. It contains AA Ser 2 - Glu 92 (Accession # NP_006263). | |
-20°C | |
S100B | |
Human | |
50 mM Tris, 150 mM NaCl, pH8.0 | |
E.coli | |
Other Recombinant Proteins |