Learn More
ACROBiosystems Human SCARB2 / SR-B2 / LIMPII Protein, Fc Tag
Marke:  ACROBiosystems SC2-H5250-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15887647
  • 1919.45 € / 1 Milligramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Scavenger receptor class B member 2 (SCARB2) is also known as Lysosome membrane protein 2 (LIMP2), 85 kDa lysosomal membrane sialoglycoprotein (LGP85), CD36 antigen-like 2 (CD36L2, LIMP2), which belongs to the CD36 family. SCARB2 acts as a lysosomal receptor for glucosylceramidase (GBA) targeting.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.6 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH21892
1 mg
Arg 27 - Thr 432
SCARB2,CD36L2,LIMP2,LIMPII
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
72.6 kDa
Human SCARB2, Fc Tag (SC2-H5250) is expressed from human 293 cells (HEK293). It contains AA Arg 27 - Thr 432 (Accession # AAH21892).
-20°C
SCARB2
Human
50 mM Tris, 100 mM Glycine, pH7.5
HEK293
Bio-Markers and CD Antigens