Learn More
ACROBiosystems Human Siglec-3 / CD33 Protein, Fc Tag
Marke:  ACROBiosystems CD3-H5257-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15886167
  • 2007.22 € / 1 Milligramm
Verfügbar ab: 02-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Myeloid cell surface antigen CD33 is also known as SIGLEC3, Siglecs (sialic acid binding Ig­like lectins) and GP67, is a single-pass type I membrane protein which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. Human CD33 / Siglec-3 cDNA encodes a 364 amino acid (aa) polypeptide with a hydrophobic signal peptide, an N-­terminal Ig-­like V­-type domain, one Ig­-like C2-­type domains, a transmembrane region and a cytoplasmic tail. CD33 / Siglec-3 usually considered myeloid-specific, but it can also be found on some lymphoid cells. In the immune response, CD33 / Siglec-3 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. CD33 / Siglec-3 induces apoptosis in acute myeloid leukemia.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 64-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH28152.1
52.9 kDa
1 mg
Asp 18 - His 259
CD33,SIGLEC3,gp67
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Human Siglec-3, Fc Tag (CD3-H5257) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).
-20°C
Siglec-3
Fc Tag
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens