Learn More
Abnova™ Human SNTB2 Partial ORF (NP_006741.1, 116 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006645-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Dystrophin is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes. [provided by RefSeq]
Sequence: VRVVKQEAGGLGISIKGGRENRMPILISKIFPGLAADQSRALRLGDAILSVNGTDLRQATHDQAVQALKRAGKEVLLEVKFIREVTPYIKKPSLVSpezifikation
NP_006741.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VRVVKQEAGGLGISIKGGRENRMPILISKIFPGLAADQSRALRLGDAILSVNGTDLRQATHDQAVQALKRAGKEVLLEVKFIREVTPYIKKPSLV | |
RUO | |
SNTB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6645 | |
SNTB2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D16S2531E/EST25263/SNT2B2/SNT3/SNTL | |
SNTB2 | |
Recombinant | |
wheat germ expression system |