Learn More
Abnova™ Human SPAG4 Partial ORF (NP_003107, 221 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006676-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The mammalian sperm flagellum contains two cytoskeletal structures associated with the axoneme: the outer dense fibers surrounding the axoneme in the midpiece and principal piece and the fibrous sheath surrounding the outer dense fibers in the principal piece of the tail. Defects in these structures are associated with abnormal tail morphology, reduced sperm motility, and infertility. In the rat, the protein encoded by this gene associates with an outer dense fiber protein via a leucine zipper motif and localizes to the microtubules of the manchette and axoneme during sperm tail development. [provided by RefSeq]
Sequence: AELDKLHKEVSTVRAANSERVAKLVFQRLNEDFVRKPDYALSSVGASIDLQKTSHDYADRNTAYFWNRFSFWNYARPPTVILEPHVFPGNCWAFEGDQGQVVIQLPGRVQSpezifikation
NP_003107 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AELDKLHKEVSTVRAANSERVAKLVFQRLNEDFVRKPDYALSSVGASIDLQKTSHDYADRNTAYFWNRFSFWNYARPPTVILEPHVFPGNCWAFEGDQGQVVIQLPGRVQ | |
RUO | |
SPAG4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6676 | |
SPAG4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPAG4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |