missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TERF2IP Full-length ORF (NP_061848.2, 1 a.a. - 399 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00054386-P01.10ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Sequence: MAEAMDLGKDPNGPTHSSTLFVRDDGSSMSFYVRPSPAKRRLSTLILHGGGTVCRVQEPGAVLLAQPGEALAEASGDFISTQYILDCVERNERLELEAYRLGPASAADTGSEAKPGALAEGAAEPEPQRHAGRIAFTDADDVAILTYVKENARSPSSVTGNALWKAMEKSSLTQHSWQSLKDRYLKHLRGQEHKYLLGDAPVSPSSQKLKRKAEEDPEAADSGEPQNKRTPDLPEEEYVKEEIQENEEAVKKMLVEATREFEEVVVDESPPDFEIHITMCDDDPPTPEEDSETQPDEEEEEEEEKVSQPEVGAAIKIIRQLMEKFNLDLSTVTQAFLKNSGELEATSAFLASGQRADGYPIWSRQDDIDLQKDDEDTREALVKKFGAQNVARRIEFRKKSpezifikation
NP_061848.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DRIP5/RAP1 | |
TERF2IP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54386 | |
TERF2IP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEAMDLGKDPNGPTHSSTLFVRDDGSSMSFYVRPSPAKRRLSTLILHGGGTVCRVQEPGAVLLAQPGEALAEASGDFISTQYILDCVERNERLELEAYRLGPASAADTGSEAKPGALAEGAAEPEPQRHAGRIAFTDADDVAILTYVKENARSPSSVTGNALWKAMEKSSLTQHSWQSLKDRYLKHLRGQEHKYLLGDAPVSPSSQKLKRKAEEDPEAADSGEPQNKRTPDLPEEEYVKEEIQENEEAVKKMLVEATREFEEVVVDESPPDFEIHITMCDDDPPTPEEDSETQPDEEEEEEEEKVSQPEVGAAIKIIRQLMEKFNLDLSTVTQAFLKNSGELEATSAFLASGQRADGYPIWSRQDDIDLQKDDEDTREALVKKFGAQNVARRIEFRKK | |
RUO | |
TERF2IP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |