Learn More
ACROBiosystems Human TGF-beta RII / TGFBR2 Protein, Fc Tag
387.09 € - 2661.24 €
Spezifikation
Zugriffsnummer | NP_003233 |
---|---|
Molekulargewicht | 42.1 kDa |
Quelle | Human TGFBR2, Fc Tag (TG2-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # NP_003233). |
Immunogen | Thr 23 - Asp 159 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15847757
|
ACROBiosystems
TG2-H5252-1MG |
1 mg |
2661.24 €
1 Milligramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15837757
|
ACROBiosystems
TG2-H5252-100UG |
100 μg |
387.09 €
100 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
TGF-beta receptor type-2 (TGFBR2 or TGFR-2) is also known as TGF-beta type II receptor, Transforming growth factor-beta receptor type II, TbetaR-II, TGFβR2, which is a homodimer or heterohexamer, belongs to the protein kinase superfamily, TKL Ser/Thr protein kinase family and TGFB receptor subfamily. TGFR2 / TGFBR2 binds TGF-β1 / TGFB1 and TGF-β3 / TGFB3 with high affinity and TGF-β2 / TGFB2 with a much lower affinity. This type I I receptor forms a heterodimeric complex with type I receptor and is essential for signal transduction. Upon ligand binding, the TGFR2 autophosphorylates its cytoplasmic domain and subsequently phosphorylates the downstream molecules which then enter the nucleus and regulate the transcription of a subset of genes related to cell proliferation.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_003233 | |
Human TGFBR2, Fc Tag (TG2-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Asp 159 (Accession # NP_003233). | |
-20°C | |
TGF-beta RII | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
42.1 kDa | |
Thr 23 - Asp 159 | |
TGFBR2,TGFR2,TbetaR-II,TGFβR2 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |