Learn More
ACROBiosystems Human TIGIT Protein, Fc Tag
MALS verified
Marke: ACROBiosystems TIT-H5254-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
T-cell immunoreceptor with Ig and ITIM domains (TIGIT) is also known as V-set and immunoglobulin domain-containing protein 9 (VSIG9), V-set and transmembrane domain-containing protein 3 (VSTM3),which belongs to single-pass type I membrane protein containing an immunoglobulin variable domain, a transmembrane domain and an immunoreceptor tyrosine-based inhibitory motif (ITIM). TIGIT is expressed at low levels on peripheral memory and regulatory CD4+ T-cells and NK cells and is up-regulated following activation of these cells (at protein level). TIGIT binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T-cell activation by promoting the generation of mature immunoregulatory dendritic cells.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 45-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_776160.2 | |
1 mg | |
Met 22 - Pro 141 | |
TIGIT,VSIG9,VSTM3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
39.7 kDa | |
Human TIGIT, Fc Tag (TIT-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accession # Q495A1-1). | |
-20°C | |
TIGIT | |
Human | |
50 mM Tris, 100 mM Glycine, 25 mM Arginine, 10 mM NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |