Learn More
R&D Systems™ Human TIMP-1 Quantikine ELISA Kit, R&D Systems™
Human TIMP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich | 0.16 - 10ng/mL |
---|---|
Assay-Empfindlichkeit: | 0.08ng/mL |
Probentyp | Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum |
Beschreibung
The Human TIMP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TIMP-1 in cell culture supernates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (10 ul).Spezifikation
0.16 - 10ng/mL | |
0.08ng/mL | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum |