missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total PON1 DuoSet IC, R&D Systems™
Human Total PON1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Kreuzreaktivität | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
---|---|
Produkttyp | ELISA Development Kit for intracellular proteins |
Menge | 1 Kit |
Gen-ID (Entrez) | 5444 |
Spezies | Human |
Beschreibung
The Human Total PON1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human PON1.Spezifikation
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
1 Kit | |
Human |
ELISA Development Kit for intracellular proteins | |
5444 |