Learn More
Abnova™ Human TRADD Partial ORF (NP_003780, 203 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008717-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a death domain containing adaptor molecule that interacts with TNFRSF1A/TNFR1 and mediates programmed cell death signaling and NF-kappaB activation. This protein binds adaptor protein TRAF2, reduces the recruitment of inhibitor-of-apoptosis proteins (IAPs) by TRAF2, and thus suppresses TRAF2 mediated apoptosis. This protein can also interact with receptor TNFRSF6/FAS and adaptor protein FADD/MORT1, and is involved in the Fas-induced cell death pathway. [provided by RefSeq]
Sequence: LFQGQPVVNRPLSLKDQQTFARSVGLKWRKVGRSLQRGCRALRDPALDSLAYEYEREGLYEQAFQLLRRFVQAEGRRATLQRLVEALEENELTSLAEDLLGLTDPNGGLASpezifikation
NP_003780 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LFQGQPVVNRPLSLKDQQTFARSVGLKWRKVGRSLQRGCRALRDPALDSLAYEYEREGLYEQAFQLLRRFVQAEGRRATLQRLVEALEENELTSLAEDLLGLTDPNGGLA | |
RUO | |
TRADD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8717 | |
TRADD (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Hs.89862/MGC11078 | |
TRADD | |
Recombinant | |
wheat germ expression system |