Learn More
ACROBiosystems Human TRAIL R2 / DR5 / TNFRSF10B Protein, Fc Tag
Marke:  ACROBiosystems TR2-H5255-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15887857
  • 2007.22 € / 1 Milligramm
Verfügbar ab: 02-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Tumor necrosis factor receptor superfamily member 10B (TNFRSF10B) is also known as TNF-related apoptosis-inducing ligand receptor 2 (TRAILR2), Death receptor 5 (DR5), CD262, KILLER, is a member of the TNF-receptor superfamily, and contains an intracellular death domain. TNFRSF10B / DR-5 is widely expressed in adult and fetal tissues; very highly expressed in tumor cell lines. TRAILR2 / CD262 / DR5 is the receptor for the cytotoxic ligand TNFSF10/TRAIL. The adapter molecule FADD (a death domain containing adaptor protein) of TRAIL-R2 / TNFRSF10B recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. CD262 / DR5 Promotes the activation of NF-kappa-B. DR5 is essential for ER stress-induced apoptosis and is regulated by p53/TP53.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_003833
40.4 kDa
1 mg
Ile 56 - Glu 182
TNFRSF10B,TRAILR2,TRAIL-R2,CD262,DR5,KILLER,TRICK2,ZTNFR9,TRICKB
Unconjugated
50 mM Tris, 100 mM NaCl, pH7.5
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Human TRAIL R2, Fc Tag (TR2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession # NP_003833).
-20°C
TRAIL R2
Fc Tag
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens