missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TTC30B Full-length ORF (AAH33795.1, 1 a.a. - 665 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00150737-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Description
Sequence: MAGLSGAQIPDGEFTAVVYRLIRNARYAEAVQLLGGELQRSPRSRAGLSLLGYCYYRLQEFALAAECYEQLGQLHPELEQYRLYQAQALYKACLYAEATRVAFLLLDNPAYHSRVLHLQAAIKYSEGDLPGSRSLVEQLPSREGGEESGGENETDGQINLGCLLYKEGQYEAACSKFFAALQASGYQPDLSYNLALAYYSSRQYASALKHIAEIIERGIRQHPELGVGMTTEGIDVRSVGNTLVLHQTALVEAFNLKAAIEYQLRNYEAAQEALTDMPPRAEEELDPVTLHNQALMNMDARPTEGFEKLQFLLQQNPFPPETFGNLLLLYCKYEYFDLAADVLAENAHLIYKFLTPYLYDFLDAVITCQTAPEEAFIKLDGLAGMLTEVLRKLTIQVQEARHNRDDEAIKKAVNEYDETMEKYIPVLMAQAKIYWNLENYPMVEKIFRKSVEFCNDHDVWKLNVAHVLFMQENKYKEAIGFYEPIVKKHYDNILNVSAIVLANLCVSYIMTSQNEEAEELMRKIEKEEEQLSYDDPDKKMYHLCIVNLVIGTLYCAKGNYDFGISRVIKSLEPYNKKLGTDTWYYAKRCFLSLLENMSKHTIMLRDSVIQECVQFLEHCELHGRNIPAVIEQPLEEERMHVGKNTVTYESRQLKALIYEIIGWNISpecifications
AAH33795.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
102.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ30990 | |
TTC30B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
150737 | |
TTC30B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGLSGAQIPDGEFTAVVYRLIRNARYAEAVQLLGGELQRSPRSRAGLSLLGYCYYRLQEFALAAECYEQLGQLHPELEQYRLYQAQALYKACLYAEATRVAFLLLDNPAYHSRVLHLQAAIKYSEGDLPGSRSLVEQLPSREGGEESGGENETDGQINLGCLLYKEGQYEAACSKFFAALQASGYQPDLSYNLALAYYSSRQYASALKHIAEIIERGIRQHPELGVGMTTEGIDVRSVGNTLVLHQTALVEAFNLKAAIEYQLRNYEAAQEALTDMPPRAEEELDPVTLHNQALMNMDARPTEGFEKLQFLLQQNPFPPETFGNLLLLYCKYEYFDLAADVLAENAHLIYKFLTPYLYDFLDAVITCQTAPEEAFIKLDGLAGMLTEVLRKLTIQVQEARHNRDDEAIKKAVNEYDETMEKYIPVLMAQAKIYWNLENYPMVEKIFRKSVEFCNDHDVWKLNVAHVLFMQENKYKEAIGFYEPIVKKHYDNILNVSAIVLANLCVSYIMTSQNEEAEELMRKIEKEEEQLSYDDPDKKMYHLCIVNLVIGTLYCAKGNYDFGISRVIKSLEPYNKKLGTDTWYYAKRCFLSLLENMSKHTIMLRDSVIQECVQFLEHCELHGRNIPAVIEQPLEEERMHVGKNTVTYESRQLKALIYEIIGWNI | |
RUO | |
TTC30B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |