Learn More
ACROBiosystems Human VEGF R3 / FLT4 Protein, Fc Tag
Marke: ACROBiosystems FL4-H5251-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Vascular endothelial growth factor receptor 3 (VEGF R3), also known as FLT-4, together with the other two members VEGFR1 (FLT-1) and VEGFR2 (KDR/Flk-1) are receptors for vascular endothelial growth factors (VEGF) and belong to the class III subfamily of receptor tyrosine kinases (RTKs). VEGF R3 mediates lymphangiogenesis in response to VEGF-C and VEGF-D. VEGF R3 is widely expressed in the early embryo but becomes restricted to lymphatic endothelia at later stages of development. It is likely that VEGF R3 may be important for lymph angiogenesis.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 111.2 kDa. As a result of glycosylation, the protein migrates as 140-160 kDa under reducing (R) condition, and 280-320 kDa under non-reducing (NR) condition (SDSpezifikation
NP_002011.2 | |
100 μg | |
Tyr 25 - Ile 776 | |
Flt-4,FLT4,LMPH1A,PCLFLT41,VEGFR3,VEGFR-3,FLT-4,FLT-41,FLT41,PCL | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
111.2 kDa | |
Human VEGF R3, Fc Tag (FL4-H5251) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Accession # NP_002011.2). | |
-20°C | |
VEGF R3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |