Learn More
ACROBiosystems Human Vitronectin / VTN Protein, Tag Free
Marke: ACROBiosystems VIN-H5119-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Vitronectin is also known as S-protein, VN, VTN, V75. Vitronectin, a multifunctional glycoprotein, is involved in coagulation, inhibition of the formation of the membrane attack complex (MAC), cell adhesion and migration, wound healing, and tissue remodeling. The primary cellular source of vitronectin is hepatocytes. Blocking of Hic(a member of the pneumococcal surface protein C (PspC) family) by specific antiserum or genetic deletion significantly reduced pneumococcal binding to soluble and immobilised vitronectin and to Factor H, respectively. In addition, Vitronectin interact with glycosaminoglycans and proteoglycans. Is recognized by certain members of the integrin family and serves as a cell-to-substrate adhesion molecule. Inhibitor of the membrane-damaging effect of the terminal cytolytic complement pathway.
This protein carries no "e;tag"e;. The protein has a calculated MW of 43.2 kDa. The protein migrates as 48-50 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_000629.3 | |
1 mg | |
Asp 20 -Arg 398 | |
VTN,V75,VN,Vitronectin,S-protein,Serum-spreading factor | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
43.2 kDa | |
Human Vitronectin, Tag Free (VIN-H5119) is expressed from E.coli cells. It contains AA Asp 20 -Arg 398 (Accession # P04004-1). | |
-20°C | |
Vitronectin | |
Human | |
20 mM Tris-HCl, Arginine, pH8.0 | |
E.coli | |
Other Recombinant Proteins |