missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WDR59 Full-length ORF (AAH04519.1, 1 a.a. - 571 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00079726-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MAARWSSENVVVEFRDSQATAMSVDCLGQHAVLSGRRFLYIVNLDAPFEGHRKISRQSKWDIGAVQWNPHDSFAHYFAASSNQRVDLYKWKDGSGEVGTTLQGHTRVISDLDWAVFEPDLLVTSSVDTYIYIWDIKDTRKPTVALSAVAGASQVKWNKKNANCLATSHDGDVRIWDKRKPSTAVEYLAAHLSKIHGLDWHPDSEHILATSSQDNSVKFWDYRQPRKYLNILPCQVPVWKARYTPFSNGLVTVMVPQLRRENSLLLWNVFDLNTPVHTFVGHDDVVLEFQWRKQKEGSKDYQLVTWSRDQTLRMWRVDSQMQRLCANDILDGVDEFIESISLLPEPEKTLHTEDTDHQHTASHGEEEALKEDPPRNLLEERKSDQLGLPQTLQQEFSLINVQIRNVNVEMDAADRSCTVSVHCSNHRVKMLVKFPAQYPNNAAPSFQFINPTTITSTMKAKLLKILKDTALQKVKRGQSCLEPCLRQLVSCLESFVNQEDSASSNPFALPNSVTPPLPTFARVTTAYGSYQDANIPFPRTSGARFCGAGYLVYFTRPMTMHRAVSPTEPTPRSpezifikation
AAH04519.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12270/FP977/MGC11230 | |
WDR59 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79726 | |
WDR59 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAARWSSENVVVEFRDSQATAMSVDCLGQHAVLSGRRFLYIVNLDAPFEGHRKISRQSKWDIGAVQWNPHDSFAHYFAASSNQRVDLYKWKDGSGEVGTTLQGHTRVISDLDWAVFEPDLLVTSSVDTYIYIWDIKDTRKPTVALSAVAGASQVKWNKKNANCLATSHDGDVRIWDKRKPSTAVEYLAAHLSKIHGLDWHPDSEHILATSSQDNSVKFWDYRQPRKYLNILPCQVPVWKARYTPFSNGLVTVMVPQLRRENSLLLWNVFDLNTPVHTFVGHDDVVLEFQWRKQKEGSKDYQLVTWSRDQTLRMWRVDSQMQRLCANDILDGVDEFIESISLLPEPEKTLHTEDTDHQHTASHGEEEALKEDPPRNLLEERKSDQLGLPQTLQQEFSLINVQIRNVNVEMDAADRSCTVSVHCSNHRVKMLVKFPAQYPNNAAPSFQFINPTTITSTMKAKLLKILKDTALQKVKRGQSCLEPCLRQLVSCLESFVNQEDSASSNPFALPNSVTPPLPTFARVTTAYGSYQDANIPFPRTSGARFCGAGYLVYFTRPMTMHRAVSPTEPTPR | |
RUO | |
WDR59 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |