missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human XG Full-length ORF (AAI00768.1, 1 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00007499-P01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16171892

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 21-06-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes the XG blood group antigen, and is located at the pseudoautosomal boundary on the short (p) arm of chromosome X. The three 5' exons reside in the pseudoautosomal region and the remaining exons within the X-specific end. A truncated copy of this gene is found on the Y chromosome at the pseudoautosomal boundary. It is transcribed, but not expected to make a Y-chromosome specific gene product. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MESWWGLPCLAFLCFLMHARGQRDFDLADALDDPEPTKKPNSDIYPKPKPPYYPQPENPDSGGNIYPRPKPRPQPQPGNSGNSGGSYFNDVDRDDGRYPPRPRPRPPAGGGGGGYSSYGNSDNTHGGDHHSTYGNPEGNMVAKIVSPIVSVVVVTLLGAAASYFKLNNRRNCFRTHEPENV
Spezifikation

Spezifikation

AAI00768.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC118758/MGC118759/MGC118760/MGC118761/PBDX
XG
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7499
XG (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MESWWGLPCLAFLCFLMHARGQRDFDLADALDDPEPTKKPNSDIYPKPKPPYYPQPENPDSGGNIYPRPKPRPQPQPGNSGNSGGSYFNDVDRDDGRYPPRPRPRPPAGGGGGGYSSYGNSDNTHGGDHHSTYGNPEGNMVAKIVSPIVSVVVVTLLGAAASYFKLNNRRNCFRTHEPENV
RUO
XG
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote