Learn More
STEMCELL Technologies IBMX
Beschreibung
- IBMX is an inhibitor of cyclic nucleotide phosphodiesterases (PDEs; IC50 = 19, 50, 18, 13, 32, 7, and 50 μM for PDE1, PDE2, PDE3, PDE4, PDE5, PDE7, and PDE11, respectively)
- By inhibiting PDEs, IBMX increases cellular cAMP and cGMP levels, activating cyclic-nucleotide-regulated protein kinases
Spezifikation
Spezifikation
| CAS | 28822-58-4 |
| Synonym | 1-Methyl-3-Isobutylxanthine; Isobutylmethylxanthin; Isobutyl methylxanthine; NSC 165960 |
| Menge | 100 mg |
| Summenformel | C10H14N4O2 |
| Reinheit | ≥ 98% |