Learn More
ICOS (CD278) Armenian Hamster, BV421, Clone: C398.4A, BD Horizon™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 565887
Additional Details : Netto-Gewicht : 0.01000kg
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat, Rhesus Ma samples. It is validated for Flow cytometry.
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.Specifications
ICOS (CD278) | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Armenian Hamster | |
25 μg | |
Cell Biology | |
Human, Mouse, Rat, Rhesus Monkey | |
Purified |
Flow Cytometry | |
C398.4A | |
Brilliant Violet 421 | |
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115 | |
Mouse D10.G4.1 T-cell clone | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG |
For Research Use Only.