Learn More
ICOS (CD278) Armenian Hamster, BV421, Clone: C398.4A, BD Horizon™
Armenian Hamster Monoclonal Antibody
Marke: BD Biosciences 565886
Weitere Details : Netto-Gewicht : 0.00840kg
Beschreibung
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat, Rhesus Ma samples. It is validated for Flow cytometry.
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat, Rhesus Ma samples. It is validated for Flow cytometry.Spezifikation
ICOS (CD278) | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Armenian Hamster | |
0.1 mg | |
Cell Biology | |
Human, Mouse, Rat, Rhesus Monkey | |
Purified |
Flow Cytometry | |
C398.4A | |
Brilliant Violet 421 | |
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115 | |
Mouse D10.G4.1 T-cell clone | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG |
For Research Use Only.