missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-γ Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: B27, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566394
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
IFN-γ Monoclonal antibody specifically detects IFN-γ in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Spezifikation
IFN-γ | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
B27 | |
Mouse | |
RUO | |
Human, Baboon, Cynomolgus, Rhesus Macaque | |
Purified |